Drukuj
Chusteczki nasączone
Dostawca: ESSENTRA POROUS TECHNOLOGIES
6-LS964-685
EPTA6-LS964-685EA 2830 PLN
EPTA6-LS964-685
Chusteczki nasączone
Chusteczki Chusteczki i ręczniki jednorazowe Chusteczki nasączone
These polyester/cellulose wipes are pre-saturated with 96% IPA and 4% DIW. They are ideal for general and manufacturing applications, as well as for industrial cleaning.
  • Perforated roll pre-wetted with alcohol
  • Canister with spring loaded lid prevents dying out and facilities dispensing
  • Alcohol saturates and evaporates quickly
  • Made in the USA
Uwaga: *Produkt(y) na tej stronie nie są objęte zakresem dyrektywy UE 2019/904