Drukuj
Pneumatycznie sterowane zawory membranowe, Integra®
Dostawca: ENTEGRIS
202-128-01
ENTE202-128-01EA 0 PLN
ENTE202-128-01
Pneumatycznie sterowane zawory membranowe, Integra®
Zawory Mikrozawory
Available compliant to SEMI Standard F57-0312, the specification for polymer components used in ultrapure water and liquid chemical distribution systems. Valve design allows ultrapure water to continuously bypass the diaphragm, eliminating water stagnation and bacteria formation in the valve. Manually adjustable, integral needle valve provides precise control of bypass flow.
  • All wetted parts are PFA and PTFE
  • No external metal components
  • Unique top loading design allows for field service
  • Espy® sensor can be added for electronic monitoring of valve
Więcej informacji o produkcie
Zobacz ceny
SPECYFIKACJE
Environmentally Preferable