Drukuj
Przeglądarka światła białego, oświetlacz LabLite, ViewOne
Dostawca: EMBITEC
VL-1000 VL-1200
EMBIVL-1000EA 1160 PLN
EMBIVL-1000 EMBIVL-1200
Przeglądarka światła białego, oświetlacz LabLite, ViewOne
Transiluminatory Transiluminatory LED
Compact benchtop illuminator helps to visually identify a specific sample well, complete lanes or rows, to aid in loading 96-well plates.
  • Uniform background for enhanced visualization of colorimetric samples
  • Waterproof top allows for visualizing wet samples such as SDS-PAGE gels or dissection samples
  • Portable and space saving
The ViewOne™ LabLite is a stand-alone bench top solution for visualizing colorimetric sample against a uniform white light background. Labs have used the ViewOne with ELISA plates, colony picking and bacterial plates,Więcej informacji o produkcie