Drukuj
Ready-to-Load™ DNA Electrophoresis, Genetic Flightplan
Dostawca: EDVOTEK
921
EDVO921EA 495 PLN
EDVO921
Ready-to-Load™ DNA Electrophoresis, Genetic Flightplan
Systemy do elektroforezy Systemy do elektroforezy poziomej
The objective of this experiment is to investigate the genetic basis of beak size variation in Galapagos finches before and after a drought event using Restriction Fragment Length Polymorphism (RFLP) analysis.
  • Introduces PCR-RFLP analysis to analyse specific nucleotide sequences
  • Understand how electrophoresis separates differently-sized molecules
  • Perform agarose gel electrophoresis
  • Complete in 45 minutes
By analysing DNA samples collected from finches with different beak sizes, students determine the association between specific genetic variations andWięcej informacji o produkcie