Drukuj
Szafki podblatowe bezpieczeństwa typ 90, UTS ergo line S-5, głębokość 500 mm
Dostawca: DUPERTHAL
29-060659-503 29-060659-403 29-060659-003 29-060657-003 29-060657-403 29-060657-503
DUEH29-060659-503EA 8000 PLN
DUEH29-060659-503 DUEH29-060659-403 DUEH29-060659-003 DUEH29-060657-003 DUEH29-060657-403 DUEH29-060657-503
Szafki podblatowe bezpieczeństwa typ 90, UTS ergo line S-5, głębokość 500 mm
Meble Szafy bezpieczeństwa
These safety cabinets are fire resistant up to 90 minutes. These under bench cabinets are equipped with wing doors and/or drawers for the storage of hazardous substances in working spaces.
  • Easy moving, single-handed operation of the doors and drawers
  • All UTS ergo line models are fitted with a cylinder lock as a standard
  • Two door technologies are available: the classical on or the push-to open wing door technology (the easy moving safety doors are opened by applying a gentle push to the front)
Drawers or wing doors remain open in any position andWięcej informacji o produkcie