Drukuj
Soft Care Man Hand Soap Dispensing System
Dostawca: Diversey
1418478
DIVY1418478EA 975 PLN
DIVY1418478
Soft Care Man Hand Soap Dispensing System
Dozowniki na mydło, balsamy, kremy
Dosing dispenser for euro bottles.
  • Robust and durable dispenser
  • Easy to clean and disinfect, drip-free
  • For use with 1 L bottles
  • Suitable for use in the field
  • Healthcare
  • Integrated arm lever or as a non-contact one