Drukuj
Uprząż zabezpieczająca przed upadkiem w stylu kamizelki standardowej, Protecta®, 3M™
Dostawca: CAPITAL SAFETY
1161609 1161610 1161611 1161615 1161616 1161617 1161642 1161643 1161644
CPSD1161609EA 0 PLN
CPSD1161609 CPSD1161610 115-2663 CPSD1161615 CPSD1161616 CPSD1161617 CPSD1161642 115-2664 CPSD1161644
Uprząż zabezpieczająca przed upadkiem w stylu kamizelki standardowej, Protecta®, 3M™
Zabezpieczenia przed upadkiem Uprzęże zabezpieczające
Protecta® Standard Vest Style Fall Arrest Harness is a sleek and modern safety harness with upgraded features. It has water and stain resistant webbing which keeps it clean and dry, and protected labels that stay readable in the long term for simple and easy inspection.
  • Stain and water resistant webbing keeps the harness cleaner and dryer
  • Steel, zinc-plated black electro-deposition paint D-rings
  • Conveniently placed front D-ring helps users stay balanced and centred
  • Auto-resetting lanyard keeper provides easy parking and release of snap hooks or carabiners