Drukuj
Pokrywy HighDuster, PREMIRA® II
Dostawca: CONTEC
PRMH1001 PRMW0001
CONTPRMH1001EA 347 PLN
CONTPRMH1001 CONTPRMW0001
Pokrywy HighDuster, PREMIRA® II
Chusteczki Zestawy i wkłady do czyszczenia
Disposable Premira II Microfibre HighDuster covers reduce fibre shedding and provide effective dry and damp dusting for hard to reach areas. The microfibre fabric offers excellent pickup power and is compliant with most cleaning protocols. Compatible with the flexible foam snake duster.
  • Ideal for hard to reach places
  • Easily slips onto a variety of dusting frames
  • Disposable, clinically superior alternative to standard dust wands
  • Picks up and holds onto debris, rather that spreading it around
  • Conformable flexible foam Snake Duster reduces operator strain
Więcej informacji o produkcie