Drukuj
Lizaty, ssaków GenLysate™, mysz
Dostawca: G Biosciences
NLM-05 NLM-06 NLM-07 NLM-08 NLM-09 CFM-011 CFM-012 CFM-013 CFM-014 CFM-015 NLM-01 NLM-02 NLM-03 NLM-04 SLM-021 SLM-0210 SLM-0211 SLM-0212 SLM-022 SLM-023 SLM-024 SLM-025 SLM-026 SLM-027 SLM-028 SLM-029 SLM-041 SLM-042
NLM-05EA 1330 PLN
NLM-05 NLM-06 NLM-07 NLM-08 NLM-09 CFM-011 CFM-012 CFM-013 CFM-014 CFM-015 NLM-01 NLM-02 NLM-03 NLM-04 SLM-021 SLM-0210 SLM-0211 SLM-0212 SLM-022 SLM-023 SLM-024 SLM-025 SLM-026 SLM-027 SLM-028 SLM-029 SLM-041 SLM-042
Lizaty, ssaków GenLysate™, mysz
Komórki
GenLysates™ are total protein tissue and cell lysates that are extracted from the tissue or cell of interest. To inhibit proteolysis, the GenLysates™ are prepared in a lysis buffer containing a complete protease inhibitor cocktail. Each lysate lot is tested by electrophoresis and Western blot analysis.
GenLysate™ total protein lysates are ready to use for SDS-PAGE and Western blotting purposes. The total proteins are being extracted from whole tissue or cells following the method of Laemmli. The proteins are extracted in a denaturing buffer containing 62 mM Tris HCl and 2% SDS with ProteaseARREST™ protease inhibitor cocktail. This minimises proteolytic damage to the proteins.
GenLysate™ total protein lysates are offered as a 150 µg lyophilised protein per vial and, following reconstitution in DI water, are ready for use. GenLysate™ is also available in a wide variety of other species and tissues. Species include: Human (both normal tissue and tumour tissue lysates available), mouse, rat, rabbit, pig, plant-related, bacteria, insect, bird, frog, fish, marsupial, earthworm, and yeast.