Drukuj
Opatrunek żelowy na oparzenia/maska na twarz
Dostawca: ORKLA CARE
51011014
CEDE51011014EA 0 PLN
CEDE51011014
Opatrunek żelowy na oparzenia/maska na twarz
Artykuły pierwszej pomocy Pierwsza pomoc na oparzenia
Large burn gel dressing provides rapid cooling and effective pain relief for first and second degree burns.
  • Sterile dressing prevents injury from penetrating deeper into skin
  • Cold reduces pain and swelling
  • Can also be used as a face mask
  • Easy to use with clear instructions
Suitable for burns on larger areas such as legs, arms, or chest. For use as a face mask, simply tear off the parts of the dressing in the pre-cut holes for eyes, nose and mouth.