Drukuj
RM 5K Roller mixer
Dostawca: CAT
60207-0060
CATA60207-0060EA 3500 PLN
CATA60207-0060
RM 5K Roller mixer
Wytrząsarki Wytrząsarki kołyskowe / rotatory
Roller mixer with 5 rollers. The synchronized rotating PVC rollers provide a uniform swaying surface, perfectly suited for simultaneous gentle and homogeneous mixing of cylindrical vessels with e.g. blood or other cell suspension.
  • Wide speed range
  • Low voltage power supply
Each roll can be removed individually to provide the use of bigger cylindrical tubes or bottles (up to 5 L). The speed is continuously adjustable.