Drukuj
Skrobak wykrywalny, SteriPlast®
Dostawca: BURKLE
5378-2030
BURK5378-2030EA 189 PLN
BURK5378-2030
Skrobak wykrywalny, SteriPlast®
Skrobaczki
PS, blue. This scraper is not only coloured blue but also has a special additive in the material that makes it visible to metal detectors or X rays. The product can therefore be rejected with standard systems for checking for foreign objects.
  • Disposable
  • Sturdy and convenient
  • Produced in a class 7 cleanroom (10000)
  • Individually packaged and then sterilised by gamma rays
  • Can be detected by colour scanners, metal detectors or X-rays
Suitable for scraping off, smoothing or cleaning as well as removing residue.