Drukuj
System fuzji elektrokomórkowej i elektroporacji, ECM™ 2001+
Dostawca: BTX
45-2045 45-2046 45-2047 45-2048 45-2049
BTXI45-2045EA 209190 PLN
BTXI45-2045 BTXI45-2046 BTXI45-2047 BTXI45-2048 BTXI45-2049
System fuzji elektrokomórkowej i elektroporacji, ECM™ 2001+
Systemy do elektroporacji Elektroporatory
The ECM 2001+ is a multifunctional electrofusion and square wave electroporation generator. The ability to generate both AC and DC waves allows for fast and efficient cell fusion in hybridoma production, hybrid cell formation, and nuclear transfer applications. This system is powerful enough to yield high transfection efficiencies for cell lines and difficult to transfect cell types including stem cells and primary cells. The gentle square wave pulse also allows for high cell viability of these cell types.
  • AC waveform of 0,2 – 2,0 MHz
  • A wide range of voltages from 5 V to 3000 V
Więcej informacji o produkcie