Drukuj
Anti-FSCN1 Mouse Monoclonal Antibody (CF647) [clone: FSCN1/416]
Dostawca: Biotium
BNC470416-500 BNC470416-100
BTIUBNC470416-500EA 2860 PLN
BTIUBNC470416-500 BTIUBNC470416-100
Anti-FSCN1 Mouse Monoclonal Antibody (CF647) [clone: FSCN1/416]
Przeciwciała
Recognizes a protein of 55 kDa, which is identified as fascin-1. Its actin binding ability is regulated by phosphorylation. Antibody to fascin-1 is a very sensitive marker for Reed-Sternberg cells and variants in nodular sclerosis, mixed cellularity, and lymphocyte depletion Hodgkin's disease. It is uniformly negative in lymphoid cells, plasma cells, and myeloid cells. Fascin-1 is also expressed in dendritic cells. This marker may be helpful to distinguish between Hodgkin lymphoma and non-Hodgkin lymphoma in difficult cases. Also, the lack of expression of fascin-1 in the neoplasticWięcej informacji o produkcie