Drukuj
Komory zliczeniowe, BLAUBRAND® Fuchs-Rosenthal
Dostawca: Brand
719805 719820
BRND719805EA 329 PLN
BRND719805 BRND719820
Komory zliczeniowe, BLAUBRAND® Fuchs-Rosenthal
Szkiełka mikroskopowe i nakrywkowe Komory zliczeniowe
This counting chamber is frequently used for counting cells.
  • Ruling differs from the customary systems for blood cell counts by ist large area of 16 mm²
  • Shows 16 large squares of one mm²
  • 16 mini squares with 0,25 mm sides and an area of 0,0625 mm²