Drukuj
Łódki wagowe
Dostawca: Brand
155540 155542 155544 155546 155548 155550
BRND155540EA 208 PLN
BRND155540 BRND155542 BRND155544 BRND155546 BRND155548 BRND155550
Łódki wagowe
Łódeczki Łódeczki wagowe
Polystyrene, impact resistant, antistatic.
  • Light and flat design
  • Wall thickness approximately 0,2 mm