Drukuj
Brady® BradyJet™ 7300 Colour Label Printers
Dostawca: Brady
176884 322666 176883 322667 322659 322658 322661 322660
BRDY176884EA 12790 PLN
BRDY176884 BRDY322666 BRDY176883 BRDY322667 BRDY322659 BRDY322658 BRDY322661 BRDY322660
Brady® BradyJet™ 7300 Colour Label Printers
Drukarki specjalistyczne Label Printers
Unleash your true colours with the BradyJet™ J7300 colour label printer. Enjoy fast, industrial-grade full colour for variable print jobs from the production line to the lab.
  • Ideal for industrial labelling that changes frequently, from regulatory signage and Lean visuals to barcoding, lab ID, GHS labelling and UL/component ID
  • LabelSense™ technology enables printer, materials and inks to talk to each other for automatic part recognition, setup and adjustments
  • Dual CMY and true black (K) ink cartridges deliver vibrant colours, deep black, dark tones,
Więcej informacji o produkcie