Drukuj
Magicard™ Prima 8 Duo Printers
Dostawca: Brady
321955 321954 321956
BRDY321955EA 17830 PLN
BRDY321955 BRDY321954 BRDY321956
Magicard™ Prima 8 Duo Printers
Drukarki specjalistyczne Document Printers
The Magicard Prima 8 printer provides a compact, reliable and robust solution for instant issuance and lamination.
  • High quality double-sided colour card printing, even to the edges
  • Equipped with a range of encoding devices to write secure electronic data onto cards at the point of issue
  • Smartcard encoding enables printing and programing your smartcard in one go
  • Application: Health and hospital labeling, facility identification, safety identification, general identification
Using a retransfer printing process, images are first printed onto a thin filmWięcej informacji o produkcie