Drukuj
Magicard™ 600 Printers
Dostawca: Brady
321948 321951 321945 321946 321949 321947 321950 321952
BRDY321948EA 10370 PLN
BRDY321948 BRDY321951 BRDY321945 BRDY321946 BRDY321949 BRDY321947 BRDY321950 BRDY321952
Magicard™ 600 Printers
Drukarki specjalistyczne Document Printers
The Magicard 600 with Wi-Fi delivers faster print speeds, higher quality and optimised security features, and integrates seamlessly with existing infrastructure for secure ID data processing.
  • High print quality - The Magicard 600 offers a wide colour gamut and prints at 600×300 dpi, ideal for photo cards with super sharp text in small fonts
  • Digital Shredding™ - Once data has been used for the print job, it is rendered unreadable, helping to ensure the security of user data
Fraud prevention - The Magicard 600 offers the ability to print a personalisedWięcej informacji o produkcie