Drukuj
Osłona przeciwrozlewowa, sztywny zamek QuickBerm®, EG-5
Dostawca: Brady
198532
BRDY198532EA 6670 PLN
BRDY198532
Osłona przeciwrozlewowa, sztywny zamek QuickBerm®, EG-5
Materiały do kontroli wycieków
The extreme durable Rigid-Lock QuickBerm® provided immediate, secure deployment and spill-readiness. The built-in Rigid-Lock wall supports maximise usable space inside the berm and greatly reduce any tripping hazard on the berm perimeter.
  • Made of PVC coated fabric
  • Black in colour
The high performance material withstands a broad range of chemicals including petroleum, oils, grease, and most acids and a broad UV resistance - durable enough for extended wear.