Drukuj
Tamper Resistant Metallised Vinyl Labels for M710, M610 and M611
Dostawca: Brady
173868 173745 173726 173819
BRDY173868EA 414 PLN
BRDY173868 BRDY173745 BRDY173726 BRDY173819
Tamper Resistant Metallised Vinyl Labels for M710, M610 and M611
Etykiety
Printable, tamper-evident, metallised vinyl labels for the M610 and M611 Portable Printers.
  • The metalized vinyl (B-362) tamper evident material is ideal for rating and serial plates
  • Use caution when removing when removing from liner as material is fragile
  • Material has great fuel and oil resistance
  • Service temperature of −70 to 100 °C (−94 to 212 °F)
  • Gloss finish
Ideal for general identification, inventory and inspection labelling, and security identification applications.
A ribbon is required to print this material. Use part number M61-R6010 forWięcej informacji o produkcie