Drukuj
Dissolvable Paper Tapes for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Dostawca: Brady
170858 170723
BRDY170858EA 355 PLN
BRDY170858 BRDY170723
Dissolvable Paper Tapes for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Etykiety
Use this water dissolvable label material (B-403) for applications that require easily removable adhesive labels. Pair with a versatile Brady portable label printer to print almost anywhere.
  • Specialised - Ideal for laboratories, restaurants, work-in-process product flows and other temporary identification applications
  • Water dissolvable - Made with a tight-bonding but completely dissolvable acrylic adhesive that dissolves in warm water within 30 seconds, leaving no residue
Matte finish, abrasion-resistant, humidity-resistant, temperature-resistant,Więcej informacji o produkcie
Zobacz ceny
SPECYFIKACJE
Environmentally Preferable