Drukuj
Blokada wtyczki, Stopower
Dostawca: Brady
065673
BRDY065673EA 131 PLN
BRDY065673
Blokada wtyczki, Stopower
Kłódki i blokady bezpieczeństwa
This plug hugger is ideal for office equipment and other frequently locked out cords.
  • For use on 2 or 3 pronged polarised plugs
  • Cord cannot be plugged in with Stopower in place
  • Device cannot be removed without a key