Drukuj

Anti-GNL1 Rabbit Polyclonal Antibody (Cy7®)

Dostawca: Bioss
BS-8040R-CY7
BOSSBS-8040R-CY7EA 4520 PLN
BOSSBS-8040R-CY7
Anti-GNL1 Rabbit Polyclonal Antibody (Cy7®)
Przeciwciała
The GNL1 gene, identified in the human major histocompatibility complex class I region, shows a high degree of similarity with its mouse counterpart. The GNL1 gene is located less than 2 kb centromeric to HLA-E, in the same transcriptional orientation. GNL1 is telomeric to HLA-B and HLA-C. [provided by RefSeq, Jul 2008].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GNL1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR