Drukuj

Anti-RERE Rabbit Polyclonal Antibody (Cy5®)

Dostawca: Bioss
BS-7077R-CY5
BOSSBS-7077R-CY5EA 4520 PLN
BOSSBS-7077R-CY5
Anti-RERE Rabbit Polyclonal Antibody (Cy5®)
Przeciwciała
Plays a role as a transcriptional repressor during development. May play a role in the control of cell survival. Overexpression of RERE recruits BAX to the nucleus particularly to POD and triggers caspase-3 activation, leading to cell death.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RERE
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR