Drukuj
Anti-ADH5 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Dostawca: Bioss
BS-12448R-A350
BOSSBS-12448R-A350EA 4480 PLN
BOSSBS-12448R-A350
Anti-ADH5 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Przeciwciała
The alcohol dehydrogenase family of proteins metabolize a wide variety of substrates, including retinol, hydroxysteroids, ethanol, aliphatic alcohols and lipid peroxidation products. ADH5 (alcohol dehydrogenase 5 (class III)), also known as FDH (formaldehyde dehydrogenase), ADHX, ADH-3 or GSNOR, is a 374 amino acid cytoplasmic protein that belongs to the class III subfamily of alcohol dehydrogenases. Expressed ubiquitously, ADH5 uses iron as a cofactor to catalytically oxidize both long-chain primary alcohols and S-hydroxymethyl-glutathione, a product formed spontaneously betweenWięcej informacji o produkcie