Drukuj
Zlew S, b.safe
Dostawca: Bohlender
R1250-20
BOHLR1250-20EA 3300 PLN
BOHLR1250-20
Zlew S, b.safe
Zlewy i aramtura Umywalki
Made of static dissipative PE-EX, with sloped bottom for complete emptying.
  • With mounted grounding cable
  • Can be covered completely
To be integrated in laboratory hoods or working tables for an easy disposal of large quantities of liquid solvent wastes. Thread connection GL 25 to connect b.safe waste tubes that transport liquid solvent wastes directly in canisters.
Drain Ø 60 mm with removable sieve. Bottom drain with male thread GL 25 for direct connection to flexible b.safe waste tubes. Grounding connection at the side with mounted grounding cableWięcej informacji o produkcie
Zobacz ceny
SPECYFIKACJE
Environmentally Preferable