Drukuj
Pierścień retortowy do tygli
Dostawca: BOCHEM
5505
BOCH5505EA 1670 PLN
BOCH5505
Pierścień retortowy do tygli
Pierścienie do statywów
Crucible retort ring made of stainless steel. Ideal for clamping of crucibles for heating substances over a gas flame.
  • Oriental clamp for crucibles, which can be fixed easily by hand according to the used crucible size: 0 to 70 mm
  • 3-point-clamp: ring with 3 guide sleeves pointing to the center, the clamping rods inside the guide sleeves can be scrolled
  • Clamping rods: ceramic rods 'Degussit' (max. 2030 °C)
  • Can be combined with support stands or tripods; with the support stands the distance between the gas flame and crucible bottom can be adjusted
  • Also
Więcej informacji o produkcie