Drukuj
Anti-CD68 Mouse Monoclonal Antibody (CF405S) [clone: KP1]
Dostawca: Biotium
BNC040512-500 BNC040512-100
BNC040512-500EA 2860 PLN
BNC040512-500 BNC040512-100
Anti-CD68 Mouse Monoclonal Antibody (CF405S) [clone: KP1]
Przeciwciała
This antibody recognizes a glycoprotein of 110 kDa, which is identified as CD68. It is important for identifying macrophages in tissue sections. It stains macrophages in a wide variety of human tissues, including Kupffer cells and macrophages in the red pulp of the spleen, in lamina propria of the gut, in lung alveoli, and in bone marrow. It reacts with myeloid precursors and peripheral blood granulocytes. It also reacts with plasmacytoid T cells, which are supposed to be of monocyte/macrophage origin. It shows strong granular cytoplasmic staining of chronic and acute myeloid leukemia andWięcej informacji o produkcie