Drukuj
Strzykawka do pobierania próbek SP Bel-Art
Dostawca: Bel-Art Products, a Part of SP
F378790000
BELAF378790000EA 325 PLN
BELAF378790000
Strzykawka do pobierania próbek SP Bel-Art
Próbniki Próbniki do cieczy
Sample liquids by simply squeezing the bulb, submersing in liquid, and releasing to draw liquids into the syringe.
  • Can be inverted to contain liquid in the graduated bulb
  • Graduation in 5 ml increments
  • Reusable item can be unscrewed from bulb for easy cleaning and storage
Certyfikaty: FDA grade materials.
Uwaga: Not recommended for use with liquids whose temperature is higher than 80 °C.
*Produkt(y) na tej stronie nie są objęte zakresem dyrektywy UE 2019/904