Drukuj
Kompaktowy system do ekstrakcji 250 ml, behrotest® KEX 250 / KEX 250 F
Dostawca: BEHR
B00217732 B00217737
BEHRB00217732EA 6740 PLN
BEHRB00217732 BEHRB00217737
Kompaktowy system do ekstrakcji 250 ml, behrotest® KEX 250 / KEX 250 F
Systemy anaerobowe
behrotest® serial extraction devices are the economical and user-friendly solution for classic Soxhlet and fat extraction. With behr hydrolysis units (six sample positions), acid digestion is possible before extraction (determination of total fat content according to Weibull and Stoldt).
  • Complete with reaction vessels, extraction spacers and coolers
  • Power is individually adjustable for each sample position
  • Cooling water distribution ensures even cooling at all sample positions
  • Practical stand for holding condensers securely between extractions
  • Holders for
Więcej informacji o produkcie