Drukuj
Zestawy do eksperymentów, chromatografia powinowactwa, G-Biosciences
Dostawca: G-Biosciences
BE-417
BE-417EA 1650 PLN
BE-417
Zestawy do eksperymentów, chromatografia powinowactwa, G-Biosciences
Materiały edukacyjne Materiały edukacyjne z chemii Chromatography Learning Activities
The Affinity Chromatography kit teaches the basic principle of affinity chromatography utilising highly specific affinity columns. This lab activity involves preparation of a crude protein extract and running affinity exchange chromatography for isolation of a protein.
  • The principle of affinity chromatography
  • Immobilisation of protein on affinity columns
  • Binding and elution of proteins on affinity columns
  • Factors influencing affinity chromatography
  • Hands-on affinity chromatography lab activity
Also required: Spectrophotometer and cuvettes orWięcej informacji o produkcie