Drukuj
Igły do znieczulenia rdzeniowego, Quincke Point, BD™
Dostawca: BD Medical
405258 405248 405253 405259 405250 405257 405256 405247 405240 405252 405255
BDAM405258EA 514 PLN
BDAM405258 BDAM405248 BDAM405253 BDAM405259 BDAM405250 BDAM405257 BDAM405256 BDAM405247 BDAM405240 BDAM405252 BDAM405255
Igły do znieczulenia rdzeniowego, Quincke Point, BD™
Igły do strzykawek
BD® Quincke Spinal Needles are available in different sizes with lengths ranging from 90 to 178 mm and gauges from 18 to 27 G. The key/slot arrangement of stylet and cannula hubs on the BD® Quincke Spinal Needles facilitates proper needle bevel orientation.
  • Enhanced precision with BD® Quincke Spinal Needles that enable visualisation of cerebrospinal fluid (CSF), aiding proper needle placement
  • Fitted stylet designed to reduce tissue coring
  • Translucent window hub featuring contact clarity that helps allow visualisation of CSF
Sposób pakowania: Packaged 2Więcej informacji o produkcie