Drukuj
BD Emerald™ Syringes
Dostawca: BD Medical
307727 307736 307731 303126 303127 303128 307742
613-2943EA 77.5 PLN
613-2943 BDAM307736 BDAM307731 BDAM303126 BDAM303127 BDAM303128 BDAM307742
BD Emerald™ Syringes
Strzykawki i igły Strzykawki medyczne
Sterile, disposable, three-piece syringes, PP barrel and piston, latex-free stopper. Luer slip syringes available with or without needle. Combining high-quality performance and reduced environmental impact.
  • For general purpose use. Specialised design features benefit clinicians, patients and the environment
  • Green stopper and bold scale markings to aid in setting an accurate dose
  • Clear barrel allows a clear view of fluid
  • Retaining ring to help prevent accidental plunger rod pullout
  • Flange size and shape to provide stability and comfort during use
  • Textured
Więcej informacji o produkcie