Drukuj
CryoPlus™ Storage Systems
Dostawca: Thermo Fisher Scientific
7405
BARN7405EA 163210 PLN
BARN7405
CryoPlus™ Storage Systems
Systemy do krioprezerwacji
Increase valuable laboratory storage space with Thermo Scientific™ CryoPlus™ storage systems, which store up to 38500 vials (2,0 ml). The ideal combination of liquid nitrogen, storage reliability and microprocessor technology, the CryoPlus series LN₂ storage system provides precise and accurate control over all parameters without complicated programming.
  • 16 preset audible and visual alarm combinations provide maximum product protection
  • Automatic defogger incorporated into the manual fill index to improve visibility during retrieval
  • Liquid level sensor and