Drukuj
AESCULAP® Acculan 4 Dermatome
Dostawca: Aesculap
GA340
AESCGA340EA 46290 PLN
AESCGA340
AESCULAP® Acculan 4 Dermatome
Ostrza, skalpele i uchwyty do ostrzy Skalpele
The acculan 4 dermatome is powerful and simple to use. It combines reliability, longevity and ergonomics. A machine that will inspire you.
  • Keyless adjustment of cutting thickness and width
  • Cutting thickness: 0,2 - 1,2 mm
  • Cutting width: max. 78 mm
  • Golden marked operating elements
The titanium housing and the ECCOS storage solution acculan 4 is suitable for mechanical cleaning, even with alkaline cleaning media.
Regular servicing and preventive maintenance is essential to ensure the reliability of motor systems over a long service life. The serviceWięcej informacji o produkcie