Drukuj
Cobalt Standard Solution, 10000 mg/L Co in dil. Nitric acid for ICP-MS
Dostawca: ACTIVE MOTIF
M112.5NP.L1
ACSDM112.5NP.L1EA 1330 PLN
ACSDM112.5NP.L1
Cobalt Standard Solution, 10000 mg/L Co in dil. Nitric acid for ICP-MS
Standardy Wzorce kationowe Cobalt Standards
Co 10000 mg/L in diluted nitric acid for ICP/MS.
Shelf life on ship date: 36 months - store under normal laboratory conditions, at temperatures between 15 to 25 °C.
Certyfikaty: ISO 17034, ISO 17025, ISO 9001.