Drukuj
Cy5 Dye qPCR Calibration Plate *Optimised for ABI7500 Fast 96-Well*
Dostawca: AAT BIOQUEST
67016
AATB67016EA 637 PLN
AATB67016
Cy5 Dye qPCR Calibration Plate *Optimised for ABI7500 Fast 96-Well*
Mikropłytki Płytki do PCR
A Cy5 dye qPCR calibration plate is a type of plate used in quantitative PCR (polymerase chain reaction) experiments.
  • The qPCR calibration plate might significantly improve qPCR results with multiplexing by more accurately representing fluorescent spectra used in your real-time experiments
  • This calibration plate is ready to use without any additional preparation steps required
For most qPCR instruments, the necessary calibrations should be run at least every six months.
Uwaga: Avoid freeze-thawing plates. It is not recommended to use the platesWięcej informacji o produkcie