Drukuj
Lumiwox™ Acridinium NHS Ester
Dostawca: AAT BIOQUEST
26000
AATB26000EA 1440 PLN
AATB26000
Lumiwox™ Acridinium NHS Ester
Zestawy do oznaczeń life science Odczynniki do oznaczania białek
The acridinium NHS ester can be used to label proteins and nucleic acids.
  • Chemiluminescent labeling
  • Improved sensitivity and stronger signal
  • Tolerates hydrogen peroxide reaction
  • Suitable for various experimental surfaces
  • Multiple applications
Uwaga: For research use only.