Drukuj
MagaDye™ 613 ddCTP
Dostawca: AAT BIOQUEST
17060 17064
AATB17060EA 6600 PLN
AATB17060 AATB17064
MagaDye™ 613 ddCTP
Odczynniki do kwasów nukleinowych Odczynniki do sekwencjonowania
Sanger sequencing, also known as the chain termination method, is a technique for DNA sequencing based upon the selective incorporation of chain-terminating dideoxynucleotides (ddNTPs) by DNA polymerase.
  • In-house quality control
  • Useful for sanger sequencing applications
Uwaga: For research use only.