Drukuj
Cell Meter™ Intracellular Fluorimetric Hydrogen Peroxide Assay Kit *Green Fluorescence Optimised for Flow Cytometry*
Dostawca: AAT BIOQUEST
11506
AATB11506EA 1800 PLN
AATB11506
Cell Meter™ Intracellular Fluorimetric Hydrogen Peroxide Assay Kit *Green Fluorescence Optimised for Flow Cytometry*
Zestawy do oznaczeń life science Testy komórkowe
Hydrogen peroxide is a reactive oxygen metabolic by-product that serves as a key regulator for a number of oxidative stress-related events.
  • Robust protocol
  • High sensitivity
  • Quantifies H₂O₂ in live cells
  • Optimized for flow cytometry
Uwaga: For research use only.