Drukuj
VWR®, High temperature viscosity standards
Dostawca: VWR Chemicals
99490.260EA 611 PLN
99490.260 99491.260 99492.260 99493.260 99495.260 99496.260 99497.260 99498.260 99499.260 99500.260 99501.260 99503.260 99504.260
VWR®, High temperature viscosity standards
Standardy Wzorce wiskozymetryczne
High Temperature viscosity standards are ideal for the calibration and verification of glass capillary viscometers and other viscosity measuring equipment where operating temperature is controlled precisely.
  • Kinematic (cSt), dynamic viscosity (mPa·s) and density (g/ml) given at all temperatures
  • Standard 500 ml pack size, in tamper-evident packaging
  • Fully traceable to national standards
  • Ensures full compliance to ASTM and IP test method protocols
Certyfikaty: Certified according to ISO 17025 / ISO 17034 under UKAS accreditation.
Uwaga: The viscosity value given in the ordering table below is the nominal viscosity value - actual batch values are given in the certification documents supplied with the product and also on the label.