Wieloelementarne standardy kotrololi jakości 10 mg/l do 36 elementów w 2% w kwasie azotowym , ARISTAR® do ICP-MS
Dostawca: VWR Chemicals
36 Elements: 10 mg/L: Al, Ag, As, B, Ba, Ca, Cd, Ce, Co, Cr, Cu, Dy, Er, Eu, Fe, Gd, Ho, K, La, Li, Lu, Mg, Mn, Na, Nd, Ni, P, Pb, Rb, Se, Sm, Sr, Tl, Tm, V, Zn
Certyfikaty: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Dodatkowe informacje: Supplied with certificate of analysis.