Wieloelementarne standardy kotrololi jakości 100 mg/l do 33 elementów, ARISTAR® do ICP
Dostawca: VWR Chemicals
33 elements 100 mg/L: Al, Ag, As, B, Ba, Be, Bi, Ca, Cd, Cs, Co, Cr, Cu, Fe, In, K, Li, Mg, Mn, Mo, Na, Ni, Nb, Pb, Rb, Sb, Se, Sr, Ti, Tl, V, U, Zn.
Certyfikaty: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Dodatkowe informacje: Supplied with certificate of analysis.