Drukuj
VWR® B'Paper, papier do pomieszczeń czystych
Dostawca: VWR Collection
818-0256EA 140 PLN
818-0256 818-0261 818-0281 818-0255 818-0254 818-0253 818-0252 818-0251 818-0250 818-0249 818-0248 818-0332
VWR® B'Paper, papier do pomieszczeń czystych
Artykuły papiernicze
Formulated with coated paper, without magnesium silicate, b-paper 80 or 92 g is cut, cleaned and packed in a Class 100 (ISO 5) cleanroom.
  • Different colours available
  • Can be used with inkjet or laser printers, with copy machines, and is also suitable for note taking
  • Low ionic and particle contamination
  • Ideal for all cleanroom applications