Drukuj
Taśmy do oznaczania, odporne na temperaturę
Dostawca: PRECISION DYNAMICS TIMEM
T-11260-1 T-11260-11 T-11260-12 T-11260-13 T-11260-14 T-11260-15 T-11260-16 T-11260-17 T-11260-18 T-11260-2 T-11260-3 T-11260-4 T-11260-5 T-11260-6 T-11260-7 T-11260-8 T-11260-9 T-1260-1 T-1260-10 T-1260-11 T-1260-12 T-1260-13 T-1260-14 T-1260-15 T-1260-16 T-1260-17 T-1260-18 T-1260-2 T-1260-3 T-1260-4 T-1260-5 T-1260-6 T-1260-7 T-1260-8 T-1260-9 T-160-1 T-160-10 T-160-11 T-160-12 T-160-13 T-160-14 T-160-16 T-160-17 T-160-18 T-160-2 T-160-3 T-160-4 T-160-5 T-160-6 T-160-7 T-160-8 T-160-9 T-260-1 T-260-10 T-260-11 T-260-12 T-260-13 T-260-14 T-260-15 T-260-16 T-260-17 T-260-18 T-260-2 T-260-3 T-260-4 T-260-5 T-260-6 T-260-7 T-260-8 T-260-9 T-3460-1 T-3460-10 T-3460-11 T-3460-12 T-3460-13 T-3460-14 T-3460-15 T-3460-16 T-3460-18 T-3460-2 T-3460-3 T-3460-4 T-3460-5 T-3460-6 T-3460-7 T-3460-8 T-3460-9 T-501-1 T-501-10 T-501-11 T-501-12 T-501-13 T-501-14 T-501-18 T-501-2 T-501-3 T-501-4 T-501-5 T-501-6 T-501-7 T-501-8 T-501-9 T-5112-1 T-5112-10 T-5112-11 T-5112-12 T-5112-13 T-5112-14 T-5112-15 T-5112-16 T-5112-17 T-5112-18 T-5112-2 T-5112-3 T-5112-4 T-5112-5 T-5112-6 T-5112-7 T-5112-8 T-512-1 T-512-10 T-512-11 T-512-12 T-512-13 T-512-14 T-512-15 T-512-16 T-512-17 T-512-18 T-512-2 T-512-3 T-512-4 T-512-5 T-512-6 T-512-7 T-512-8 T-512-9 T-534-1 T-534-10 T-534-11 T-534-12 T-534-13 T-534-14 T-534-16 T-534-18 T-534-2 T-534-3 T-534-4 T-534-5 T-534-6 T-534-7 T-534-9 T-534-8 T-534-15 T-534-17 T-502-1 T-502-10 T-502-11 T-502-12 T-502-13 T-502-14 T-502-15 T-502-16 T-502-17 T-502-18 T-502-2 T-502-3 T-502-4 T-502-5 T-502-6 T-502-7 T-502-8 T-502-9
817-0313EA 364 PLN
817-0313 817-0315 817-0316 817-0317 817-0318 817-0319 817-0320 817-0321 817-0322 817-0323 817-0324 817-0325 817-0326 817-0327 817-0328 817-0329 817-0330 817-1600 817-1609 817-1610 817-1611 817-1612 817-1613 817-0045 817-0046 817-0047 817-1614 817-1601 817-1602 817-1603 817-1604 817-1605 817-1606 817-1607 817-1608 817-1630 817-1639 817-1640 817-1641 817-1642 817-1643 817-0218 817-0048 817-1644 817-1631 817-1632 817-1633 817-1634 817-1635 817-1636 817-1637 817-1638 817-0035 TIMET-260-10 817-0036 TIMET-260-12 TIMET-260-13 TIMET-260-14 817-0037 TIMET-260-16 TIMET-260-17 TIMET-260-18 817-0021 817-0022 817-0023 817-0024 TIMET-260-6 817-0025 TIMET-260-8 TIMET-260-9 817-1615 817-1624 817-1625 817-1626 817-1627 817-1628 817-0049 817-0050 817-1629 817-1616 817-1617 817-1618 817-1619 817-1620 817-1621 817-1622 817-1623 817-1541 817-1550 817-1551 817-1552 817-1553 817-1554 817-1555 817-1542 817-1543 817-1544 817-1545 817-1546 817-1547 817-1548 817-1549 817-1538 817-0359 817-0360 817-0361 817-0362 817-0363 817-0364 817-0365 817-0366 817-0367 817-1539 817-0368 817-0369 817-0370 817-1540 817-1556 817-0371 817-1501 817-1510 817-1511 817-1512 817-1513 817-1514 817-0039 817-0040 817-0041 817-1515 817-1502 817-1503 817-1504 817-1505 817-1506 817-1507 817-1508 817-1509 817-1521 817-1530 817-1531 817-1532 817-1533 817-1534 817-0043 817-1535 817-1522 817-1523 817-1524 817-1525 817-1526 817-1527 817-1529 817-1528 817-0373 817-0374 TIMET-502-1 TIMET-502-10 TIMET-502-11 TIMET-502-12 TIMET-502-13 TIMET-502-14 TIMET-502-15 TIMET-502-16 TIMET-502-17 TIMET-502-18 TIMET-502-2 TIMET-502-3 TIMET-502-4 TIMET-502-5 TIMET-502-6 TIMET-502-7 TIMET-502-8 TIMET-502-9
Taśmy do oznaczania, odporne na temperaturę
Taśmy
This all-purpose, self-adhesive tape is well suited for a wide variety of labelling applications. It can be written on with pen, pencil or permanent marker.
  • Temperature resistant from −45 to +121 °C
  • With removable adhesive
  • Oil, water, and acid resistant, but not designed to be fully submerged in water
This tape is available in various colours, in two lengths and five widths.