Drukuj
femtoCHROMO™- AP do detekcji chromogeniczej
Dostawca: G Biosciences
786-379 786-380 786-381 786-382 786-383
786-379EA 1950 PLN
786-379 786-380 786-381 786-382 786-383
femtoCHROMO™- AP do detekcji chromogeniczej
Systemy do etykietowania i łączenia białek
A ready to use modified BCIP (5-Bromo-4-Chloro-3’-Indolyphosphate p-Toluidine salt) and NBT (Nitro-Blue Tetrazolium Chloride) substrate that generates a black-purple insoluble precipitate in the presence of alkaline phosphatase.
  • Detects >5 ng
  • Ready to use, single detection step
  • High signal to background ratio and reproducibility
Dodatkowe informacje: Supplied with an enhanced blocking agent, BLOT-QuickBlocker™, and a concentrated [10X] washing buffer, femtoTBST™ Buffer, to ensure low background staining. Optional AP labelled goat α-mouse or rabbit antibodies are also supplied.
Zobacz ceny
SPECYFIKACJE
Environmentally Preferable
Gospodarz