Drukuj
Barwnik do żelu do elektroforezy, Reversible Copper Stain™.
Dostawca: G Biosciences
786-32CU 786-32DSCU
786-32CUEA 2330 PLN
786-32CU 786-32DSCU
Barwnik do żelu do elektroforezy, Reversible Copper Stain™.
Barwniki Barwniki do elektroforezy
The Reversible Copper Stain™ is a single step stain for the rapid detection of proteins resolved on SDS-PAGE.
  • Based on the interaction of copper ions with polyacrylamide and proteins
  • Copper metal precipitate in the gel resulting in an opaque blue/green gel
  • Sensitivity of the Reversible Copper Stain™ is 0,1 to 0,5 ng BSA protein
Suitable for staining of protein gels prior to transfer or electroelution of proteins.
Uwaga: Not suitable for native gels or gels containing Tricine or Glycine.
Zobacz ceny
SPECYFIKACJE
Environmentally Preferable