Fraction-FOCUS™
Supplier: G Biosciences
786-168
786-168EA 6470 PLN
786-168
Fraction-FOCUS™
Protein Extraction Protein Extraction Kits
Fraction-FOCUS™ utilises proven technology to fractionate and concentrate all proteomes into multiple fractions; simplifying 2D maps and enhancing detection of low abundant proteins. Fraction-FOCUS™ is fully compatible with all downstream protein identification techniques.
There is no detectable loss of total protein during the Fraction-FOCUS™ procedure. At the end of the fractionation, cellular proteins are in one of many fractions.
Order Now
SPECIFICATIONS
Environmentally Preferable