Drukuj
Czytnik mikropłytek, Multiskan™ FC
Dostawca: THERMO LABSYSTEMS LIFE SCIENCE
51119000 51119100
736-0355EA 33440 PLN
736-0355 736-0356
Czytnik mikropłytek, Multiskan™ FC
Czytniki mikropłytek
The Multiskan™ FC is a compact, reliable and robust microplate photometer capable of reading 96-well plates for a wide variety of applications. It provides fast and accurate measurement with excellent linearity. One model in the range is fitted with an incubator providing temperature control up to 50 °C and the capacity to read 384-well plates. It takes only 6 seconds to measure a 96-well plate and 12 seconds to measure a 384-well plate. The wavelength range of 340 to 850 nm makes the Multiskan™ FC suited to applications from enzyme kinetics to Lowry assays.
  • Easy to use multi-
Więcej informacji o produkcie